missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ H1F0 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human H1F0 (P07305) (Met 1-Lys 194) fused to a polyhistidinetag at the N-terminus

Brand:  Thermo Scientific™ 10920H07E50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15267385

  • £323.76 / 50µg


Estimated Shipment
15-03-2018
Add to basket

Description

Description

H1 Histone Family, Member 0 (H1F0), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 600mM NaCl, 1mM DTT, pH 8.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3005
PAGE
50μg
N-Terminal
<1 EU/μg
H1 histone family, member 0
Assay Standard
E. coli
Lyophilized
22.4kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
HSPD1, CPN60, GROEL, HLD4, HSP60, HSP65, HuCHA60, SPG13
H1F0
Histones
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.