Sino Biological™ GPNMB Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human GPNMB isoform 2 (Q14956-2) extracellular domain (Met 1-Pro 474) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 11305H08H50

UNSPSC: 12352200

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15218515

Quantity Price
1 £ 197.0 / 50µg
EU Stock 3
Add to basket


Description and Specification

Specification

Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Purification Method PAGE
Common Name Glycoprotein (transmembrane)nMb
Gene ID (Entrez) 10457
Expression System Mammalian
Format Lyophilized
Gene Symbol GPNMB
Protein Tag His Tag
Structural Form Truncated/Recombinant
Purity ≥90% by SDS-PAGE
Protein Family Other Proteins
Termination C-Terminal
Endotoxin Concentration <1 EU/μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Validated Application Assay Standard
Quantity 50μg
Gene Alias UNQ1725/PRO9925, HGFIN,nMB
Molecular Weight 52.1kDa

Glycoprotein (Transmembrane)nMB (GPNMB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.