missing translation for 'onlineSavingsMsg'
Learn More

GPC3 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human GPC3 (P51654-1) (Met 1-His 559) fused to a C-terminal polyhistidine tag

£58.45 - £125.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias GPC3, AC002420.1, DGSX, GTR2-2, MXR7, OCI-5, SDYS, SGB, SGBS, SGBS1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15284625 View Documents Invitrogen
10088H08H5
5μg £58.45
5µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-06-2020
Please to purchase this item. Need a web account? Register with us today!
15264625 View Documents Invitrogen
10088H08H25
5 x 5μg £125.00
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-06-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glypican 3 (GPC3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human GPC3 at 5μg/mL (100μL/well) can bind human bFGF/FGF2 with a linear range of 0.156-2.5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
Lyophilized
GPC3, AC002420.1, DGSX, GTR2-2, MXR7, OCI-5, SDYS, SGB, SGBS, SGBS1

For Research Use Only. Not for use in diagnostic procedures.