missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ GPC3 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human GPC3 (P51654-1) (Met 1-His 559) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10088H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15264625

  • £122.84 / 25µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Glypican 3 (GPC3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human GPC3 at 5μg/mL (100μL/well) can bind human bFGF/FGF2 with a linear range of 0.156-2.5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
62kDa
5 x 5μg
Proteoglycans
GPC3, AC002420.1, DGSX, GTR2-2, MXR7, OCI-5, SDYS, SGB, SGBS, SGBS1
GPC3
Cell Structure/Organelle Proteins
His Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Glypican 3
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.