missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ GAPDH Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human GAPDH (P04406) (Met 1-Glu 335) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10094H07E5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15214635

  • £82.24 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 30% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2597
PAGE
5μg
Full Length/Recombinant
Glyceraldehyde-3-Phosphate Dehydrogenase
Assay Standard
E. coli
Frozen
38kDa
≥90% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
CDABP0047, G3PD, GAPD, mgC88685
GAPDH
Other Proteins
N-His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.