missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FTH1 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human FTH1 (P02794) (Met 1-Ser183)

Brand:  Thermo Scientific™ 13217HNAE25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15246255

  • £103.06 / 25µg


Estimated Shipment
06-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Ferritin, Heavy Polypeptide 1 (FTH1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2495
PAGE
5 x 5μg
Full Length/Recombinant
<1 EU/μg
Ferritin, heavy polypeptide 1
Assay Standard
E. coli
Lyophilized
21.2kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
OK/SW-cl.84, FHC, FTH, FTHL6, PIG15, PLIF
FTH1
Other Proteins
Videos