missing translation for 'onlineSavingsMsg'
Learn More

FSTL1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human FSTL1 (NP_009016.1) precursor (Met 1-Ile 308) fused to a carboxy-terminal polyhistidine tag was expressed

Brand:  Invitrogen 10924H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15297385

  • £75.75 / 5µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Follistatin-Like Protein 1 (FSTL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
11167
PAGE
5μg
C-Terminal
Activins|BMPs (Bone Morphogenetic Proteins) & Related Factors|TGFs (Transforming Growth Factors)
FSTL1, FLJ50214, FLJ52277, FRP, FSL1
FSTL1
Other Proteins
His Tag
Lyophilized
34.2kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
Follistatin-like protein 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.