missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FN1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the Fragment 2 (Ser 607 - Pro 1265) of human FN1 (CAD91166) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10314H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15205145

  • £144.70 / 25µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Fibronectin 1 (FN1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse APCS at 10μg/mL (100μL/well) can bind biotinylated FN1 with a linear range of 0.625 - 5μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
73.2kDa
5 x 5μg
Truncated/Recombinant
<1 EU/μg
Fibronectin 1
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Fibronectin
FN, CIG, FNZ, MSF, ED-B, FINC, GFND, LETS, GFND2
FN1
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.