missing translation for 'onlineSavingsMsg'
Learn More

FLT1 (VEGFR1) Recombinant Human Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 756) of human FLT1 (P17948-1) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 10136H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 13485726

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Fms-related tyrosine kinase 1 (FLT1/VEGFR1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit the VEGF-dependent proliferation of human umbilical vein endothelial cells (HUVEC) (Conn, G. et al., 1990 , Proc. Natl. Acad. Sci. USA 87:1323); the ED50 for this effect is typically 5-30ng/mL in the presence of 5ng/mL human VEGF165
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
83.7kDa
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
fms-related tyrosine kinase 1
VEGF Receptors
Mammalian
Lyophilized
≥85% by SDS-PAGE
2321
5μg
Truncated/Recombinant
FLT, VEGFR1, FLT1
FLT1
His Tag
Growth Factors & Receptors
Receptor Tyrosine Kinases, Growth Factor Receptors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.