missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FLRT3 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human FLRT3 (NP_938205.1) extracellular domain (Met 1-Pro 528) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11166H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15298325

  • £206.12 / 50µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Fibronectin Leucine Rich Transmembrane Protein 3 (FLRT3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
23767
PAGE
50μg
C-Terminal
<1 EU/μg
Fibronectin leucine rich transmembrane protein 3
Assay Standard
Mammalian
Lyophilized
58kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
FLRT3
FLRT3
Cell Structure/Organelle Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.