missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FKBP1A (FKBP12) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human FKBP12 (NP_463460) (Met 1-Glu 108) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10268H08E250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15265045

  • £328.96 / 250µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

FK506 binding protein 1A, 12kDa (FKBP1A), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM HEPES, 0.1 M NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2280
PAGE
5 x 50μg
C-Terminal
FKBP1A, RP11-314N13.2, FKBP-12, FKBP1, FKBP12, FKBP12C, PKC12, PKCI2, PPIASE
FKBP1A
Proteases, Peptidases & Inhibitors/Phosphatase Inhibitors
His Tag
Lyophilized
12.9kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
FK506 binding protein 1A, 12kDa
Assay Standard
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.