missing translation for 'onlineSavingsMsg'
Learn More

FIGF Recombinant Mouse Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse FIGF (P97946) (Phe 98-Ser 206) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Invitrogen 50157M01H25

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13453129

  • £261.00 / 50µg

Estimated Shipment date
from Supplier 09-03-2020

Please to purchase this item. Need a web account? Register with us today!


Includes: Five vials of lyophilized protein

Description

Description

C-fos induced growth factor (vascular endothelial growth factor D) (FIGF/VEGFD), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human VEGFR3-His at 10μg/mL (100μL/well) can bind mouse Fc-VEGFD; the EC50 of mouse VEGFD-Fc is 49ng/mL; 2. Immobilized mouse VEGFR3-His at 10μg/mL (100μL/well) can bind mouse Fc-VEGFD, the EC50 of mouse VEGFD-Fc is 44ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
40.6kDa
5 x 5μg
Full Length/Recombinant
<1 EU/μg
c-fos induced growth factor
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Growth Factors
RP23-202H24.2, AI325264, VEGF-D, Vegfd
FIGF
Growth Factors & Receptors
IgG1 Fc Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.