Sino Biological™ FIGF Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human VEGFD (NP_004460.1) corresponding to amino acid (Phe 93-Ser 201) fused to a C-terminal polyhistidine tag

Overview
Brand: Invitrogen

Manufacturer Part Number: 10557H08H25

FIGF RECOMBINANT HUMAN PROTEIN, HIS TAG

UNSPSC: 12352202

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15276915

Quantity Price
1 £ 292.0 / 25µg
Estimated Shipment date
from Supplier 06-03-2017
Add to basket


Description and Specification

Specification

Molecular Weight 20kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Full Length/Recombinant
Protein Subtype VEGF (Vascular Endothelial Growth Factor)
Endotoxin Concentration <1 EU/μg
Gene Alias FIGF, VEGF-D, VEGFD
Common Name c-fos induced growth factor (vascular endothelial growth factor D)
Gene Symbol FIGF
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Growth Factors & Receptors
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 2277

C-Fos Induced Growth Factor (Vascular Endothelial Growth Factor D) (FIGF), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.