Sino Biological™ FH Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human FH (P07954-1) (Ala 45-Lys 510) fused to a polyhistidine tag at the C-terminus and an initial Met at the N-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 12115-H08E-50

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15127562

Quantity Price
1 £ 169.0 / 50µg
Estimated Shipment date
from Supplier 16-05-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 2271
Molecular Weight 52kDa
Purification Method PAGE
Purity >85% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Termination C-Terminal
Structural Form Full Length/Recombinant
Gene Alias HLRCC, LRCC, MCL, MCUL1
Common Name Fumarate Hydratase
Gene Symbol FH
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
Protein Tag His Tag

Fumarate Hydratase (FH), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 10mM Tris, 5mM EDTA, 1mM DTT, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.