missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FH Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human FH (P07954-1) (Ala 45-Lys 510) fused to a polyhistidine tag at the C-terminus and an initial Met at the N-terminus

Brand:  Thermo Scientific™ 12115H08E25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15245765

  • £115.56 / 25µg


Estimated Shipment
06-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Fumarate Hydratase (FH), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 10mM Tris, 5mM EDTA, 1mM DTT, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2271
PAGE
5 x 5μg
C-Terminal
HLRCC, LRCC, MCL, MCUL1
FH
Other Proteins
His Tag
Lyophilized
52kDa
>85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
Fumarate Hydratase
Assay Standard
E. coli
Videos