missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FGFR4, Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 369) of human FGFR4 (NP_002002.3) precursor fused to the C-terminal polyhistidine tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10538H03H250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15286895

  • £655.83 / 250µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Fibroblast Growth Factor Receptor 4 (FGFR4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit FGF acidic dependent proliferation of Balb3T3 mouse fibroblasts, the ED50 for this effect is typically 2-6ng/mL
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
66kDa
≥85% by SDS-PAGE
Store at -5° to -30°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
fibroblast growth factor receptor 4
Assay Standard
Mammalian
2264
PAGE
5 x 50μg
Truncated/Recombinant
FGFR4, CD334, JTK2, mgC20292, TKF
FGFR4
Cytokines & Receptors
hIgG1-Fc.His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.