missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FGFR2 Recombinant Human Protein (without Catalytic Activity Domain), His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 377) of human FGFR2 (NP_000132.3) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10824H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465976

  • £56.74 / 5µg


Estimated Shipment
09-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Fibroblast growth factor receptor 2 (FGFR2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in cell-based assays, protein structure analysis, as immunogen, protein standard, and other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of Balb/c 3T3 mouse embryonic fibroblasts, the ED50 for this effect is typically 200-400ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2263
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Receptor Tyrosine Kinases|Growth Factor Receptors
BEK, BFR-1, CD332, CEK3, CFD1, ECT1, FLJ98662, JWS, K-SAM, KGFR, TK14, TK25
FGFR2
Growth Factors & Receptors
FGF Receptor Kinase Family
Lyophilized
41kDa
5μg
Truncated/Recombinant
<1 EU/μg
Fibroblast growth factor receptor 2
Assay Standard
Mammalian
His Tag
Videos