missing translation for 'onlineSavingsMsg'
Learn More

FGFR2 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 377) of human FGFR2 (NP_000132.3) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

£57.90 - £870.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias BEK, BFR-1, CD332, CEK3, CFD1, ECT1, FLJ98662, JWS, K-SAM, KGFR, TK14, TK25
Gene ID (Entrez) 2263
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13425976 View Documents Invitrogen
10824H03H5
5μg £57.90
5µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-06-2020
Please to purchase this item. Need a web account? Register with us today!
13405976 View Documents Invitrogen
10824H03H25
5 x 5μg £148.00
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-06-2020
Please to purchase this item. Need a web account? Register with us today!
13435976 View Documents Invitrogen
10824H03H50
50μg £240.00
50µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-06-2020
Please to purchase this item. Need a web account? Register with us today!
13415976 View Documents Invitrogen
10824H03H250
5 x 50μg £870.00
250µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-06-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fibroblast growth factor receptor 2 (FGFR2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BALB/3T3 mouse fibroblasts, the ED50 for this effect is typically 3.7ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
2263
<1 EU/μg
BEK, BFR-1, CD332, CEK3, CFD1, ECT1, FLJ98662, JWS, K-SAM, KGFR, TK14, TK25

For Research Use Only. Not for use in diagnostic procedures.