missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FGFR1 Recombinant Human Protein (without Catalytic Activity Domain), His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 285) of human FGFR1 (NP_075594.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10616H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13485916

  • £141.58 / 25µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Fibroblast growth factor receptor 1 (FGFR1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BALB/c 3T3 mouse fibroblasts, the ED50 for this effect is typically 8-40ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2260
>98% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Receptor Tyrosine Kinases|Growth Factor Receptors
FGFR1, BFGFR, CD331, CEK, FGFBR, FLG, FLJ99988, FLT2, HBGFR, KAL2, N-SAM
FGFR1
Growth Factors & Receptors
FGF Receptor Kinase Family
Lyophilized
31kDa
5 x 5μg
Truncated/Recombinant
<1 EU/μg
Fibroblast growth factor receptor 1
Assay Standard
Mammalian
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.