missing translation for 'onlineSavingsMsg'
Learn More

FGF10 Recombinant Human Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mature form of human FGF10 (NP_004456.1) (Gln 38-Ser 208)

Brand:  Invitrogen 10573HNAE50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15236955

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Fibroblast Growth Factor 10 (FGF10), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
19.6kDa
50μg
Full Length/Recombinant
Fibroblast Growth Factor 10
Untagged
Growth Factors & Receptors
≥90% by SDS-PAGE
2255
PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
FGF10
FGF10
E. coli
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.