missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FGF1 (aFGF) Recombinant Mouse Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form (Phe 16-Asp 155) of mouse FGF1 (NP_034327.1)

Brand:  Thermo Scientific™ 50179MNAE5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13413139

  • £56.30 / 5µg


Estimated Shipment
12-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Fibroblast growth factor 1 (FGF1/aFGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard or in other research applications.
  • Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse fibroblasts; the ED50 for this effect is typically 0.05-0.25ng/mL in the presence of 10μg/mL heparin
  • Formulation: Lyophilized in 20mM MES buffer, pH 6.0, 200mM NaCl, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
14164
>98% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
FGFs (Fibroblast Growth Factors)
Fibroblast growth factor 1
Assay Standard
E. coli
Lyophilized
16kDa
5μg
Recombinant
Dffrx, Fam, Fgf-1, Fgfa, FGF1
FGF1
Growth Factors & Receptors
Untagged
Videos