missing translation for 'onlineSavingsMsg'
Learn More

FCGR4 (CD16-2) Recombinant Mouse Protein, His.AVI Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 203) of mouse FCGR4 (NP_653142.2) fused to polyhistidine and AVI tags at the C-terminus

Brand:  Invitrogen 50036M27H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465626

  • £216.62 / 50µg


Estimated Shipment
10-04-2019
Add to basket

Description

Description

Fc gamma receptor IV (FCGR4/CD16-2), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
246256
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins|Fc Receptors
4833442P21Rik, CD16-2, FcgRIV, FcgammaRIV, FCGR3a, Fcrl3, FCGR4
FCGR4
CD Proteins
AVI Tag, His Tag
Lyophilized
24.2kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Fc receptor, IgG, low affinity IV
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.