missing translation for 'onlineSavingsMsg'
Learn More

FCGR3A (CD16a), 176 Phe/Val Recombinant Human Protein, His.AVI Tag, Invitrogen™ Sino Biological™, Biotinylated

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 208) of human FCGR3A (P08637-1), containing the 176 Phe/Val mutation, fused to a polyhistidine and AVI tags at the C-terminus

Brand:  Invitrogen 10389H27H1B5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13445866

  • £56.55 / 5µg

Estimated Shipment date
from Supplier 05-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fc gamma receptor III-a (FCGR3A/CD16a), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard. This protein is biotinylated.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized Protein A at 0.5μg/well can bind human IgG1 with a linear range of 0.078-0.625μg/mL with 3mg/mL of biotinylated human FCGR3A
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2214
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins|Fc Receptors
RP11-5K23.1, CD16, CD16A, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
FCGR3A
Growth Factors & Receptors/CD Proteins
AVI Tag, His Tag
Lyophilized
25.6kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Fc fragment of IgG, low affinity IIIa, receptor (CD16a)
Assay Standard
Mammalian
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.