missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FCGR2A (CD32A) Recombinant Human Protein, His.AVI Tag, Biotinylated

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Met 210) of human FCGR2A (AAA35827.1) (167Arg/His) fused to a C-terminal polyhistidine tagged AVI tag at the C-terminus

Brand:  Thermo Scientific™ 10374H27H1B25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15215285

  • £241.52 / 25µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Fc Gamma Receptor II-A (FCGR2A), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme)
  • Activity: 1.Measured by its binding ability in a functional ELISA; immobilized human IgG2 at 10μg/mL (100μL/well) can bind biotinylated human FCGR2A with a linear range of 0.16-0.8μg/mL
  • 2. Using the Octet RED System, the recombinant human FCGR2A and BirA bound to human IgG; the affinity constant (Kd) was 12nM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
23.6kDa
5 x 5μg
Fc Receptors
RP11-5K23.6, CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A, FCGR2A1, FcGR, IGFR2, mgC23887
FCGR2A
CD Proteins
AVI Tag, His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Fc gamma receptor II-a
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.