missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FCGR2A (CD32a) Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 218) of human FCGR2A (AAA35827.1) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10374H02H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13495836

  • £801.57 / 250µg

Estimated Shipment
Add to basket



Fc gamma receptor II-a (FCGR2A/CD32a), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins|Fc Receptors
RP11-5K23.6, CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A, FCGR2A1, IGFR2, mgC23887, mgC30032
CD Proteins
IgG1 Fc Tag
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Fc fragment of IgG, low affinity IIa, receptor (CD32)
Assay Standard

For Research Use Only. Not for use in diagnostic procedures.