missing translation for 'onlineSavingsMsg'
Learn More

FCGR2A (CD32a), 167 Arg/His Recombinant Human Protein, His.AVI Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Met 210) of human FCGR2A (AAA35827.1), containing the 167 Arg/His mutation, fused to polyhistidine and AVI tags at the C-terminus

£58.00 - £820.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-5K23.6, CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A, FCGR2A1, IGFR2, mgC23887, mgC30032
Gene ID (Entrez) 2212
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13425856 View Documents Invitrogen
10374H27H15
5μg £58.00
5µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 17-06-2020
Please to purchase this item. Need a web account? Register with us today!
13495846 View Documents Invitrogen
10374H27H125
5 x 5μg £147.00
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 17-06-2020
Please to purchase this item. Need a web account? Register with us today!
13435856 View Documents Invitrogen
10374H27H150
50μg £227.00
50µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 17-06-2020
Please to purchase this item. Need a web account? Register with us today!
13415856 View Documents Invitrogen
10374H27H1250
5 x 50μg £820.00
250µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 17-06-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fc gamma receptor II-a (FCGR2A/CD32a), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
2212
<1 EU/μg
RP11-5K23.6, CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A, FCGR2A1, IGFR2, mgC23887, mgC30032

For Research Use Only. Not for use in diagnostic procedures.