missing translation for 'onlineSavingsMsg'
Learn More

FCGR2A (CD32a), 167 Arg/His Recombinant Human Protein, His.AVI Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Met 210) of human FCGR2A (AAA35827.1), containing the 167 Arg/His mutation, fused to polyhistidine and AVI tags at the C-terminus

Brand:  Invitrogen 10374H27H150

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13435856

  • £227.00 / 50µg

Estimated Shipment
23-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fc gamma receptor II-a (FCGR2A/CD32a), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Glycoproteins|Fc Receptors
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2212
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Fc fragment of IgG, low affinity IIa, receptor (CD32)
AVI Tag, His Tag
CD Proteins
Assay Standard
Lyophilized
23.6kDa
50μg
Truncated/Recombinant/Cell Surface Protein
RP11-5K23.6, CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A, FCGR2A1, IGFR2, mgC23887, mgC30032
FCGR2A
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.