missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FCGR1A (CD64) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 288) of human FCGR1A (NP_000557.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10256H08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13472949

  • £839.05 / 250µg

Estimated Shipment
Add to basket



Fc gamma receptor I-a (FCGR1A/CD64), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized CD64 at 10μg/mL can bind human IgG1 with a linear range of 0.39-25ng/mL; 2. Scatchard analysis showed that the affinity constant (Kd) of CD64 bound to human IgG1 was 0.059nM
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
>85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins|Fc Receptors
FCGR1A, CD64, CD64A, FCRI, FLJ18345, IGFR1
Growth Factors & Receptors/CD Proteins
His Tag
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Fc fragment of IgG, high affinity Ia, receptor (CD64)
Assay Standard

For Research Use Only. Not for use in diagnostic procedures.