missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FCGR1 (CD64) Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 297) of mouse FCGR1 precursor (NP_034316.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50086M08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13416206

  • £140.54 / 25µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Fc gamma receptor I (FCGR1/CD64), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
14129
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins|Fc Receptors
FCGR1, AI323638, AV092959, CD64, FcgammaRI, IGGHAFC
FCGR1
Growth Factors & Receptors/CD Proteins
His Tag
Lyophilized
32kDa
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Fc receptor, IgG, high affinity I
Assay Standard
Mammalian
Videos