missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FCER2 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CD23 isoform 2 (P06734-1) extracellular domain (Asp 48-Ser 321) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10261H07H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15275035

  • £737.03 / 250µg


Estimated Shipment
06-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Fc Fragment of IgE, low affinity II receptor (FCER2/CD23), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2208
PAGE
5 x 50μg
N-Terminal
<1 EU/μg
Fc fragment of IgE, low affinity II, receptor for (CD23)
Assay Standard
Mammalian
Lyophilized
33.2kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
CLEC-6, CLEC6, CLECSF8, MCL, mgC40078, MPCL
FCER2
CD Proteins
His Tag
Videos