missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FAS (CD95) Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 169) of mouse FAS (NP_032013.2) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50027M08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13425616

  • £799.49 / 250µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

Fas (TNF receptor superfamily, member 6), (CD95), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
14102
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
TNF Receptors
Fas, AI196731, APO1, APT1, CD95, TNFR6, Tnfrsf6, lpr
FAS
Growth Factors & Receptors/CD Proteins
His Tag
Lyophilized
18kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Fas (TNF receptor superfamily member 6)
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.