missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FAM3D Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human FAM3D (Q96BQ1-1) (Met 1-Phe 224) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11659H08H5

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15238805

  • £93.43 / 5µg


Estimated Shipment
09-07-2018
Add to basket

Description

Description

Family with sequence similarity 3, member D (FAM3D), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
  • This protein is manufactured by Sino Biological Inc.

    Specifications

    Specifications

    Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    131177
    PAGE
    5μg
    C-Terminal
    Other Proteins
    UNQ567/PRO1130, EF7, OIT1
    FAM3D
    Other Proteins
    His Tag
    Lyophilized
    23.4kDa
    ≥90% by SDS-PAGE
    Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Full Length/Recombinant
    <1 EU/μg
    Family with sequence similarity 3, member D
    Assay Standard
    Mammalian
    Videos

    For Research Use Only. Not for use in diagnostic procedures.