missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ FABP6 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human FABP6 (AAH22489.1) (Met 1 -Ala 128)

Brand:  Thermo Scientific™ 12579HNAE50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15225995

  • £213.41 / 50µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Fatty Acid Binding Protein 6, ileal (FABP6), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2172
PAGE
50μg
Full Length/Recombinant
I-15P, I-BABP, I-BALB, I-BAP, ILBP, ILBP3, ILLBP
FABP6
Other Proteins
Lyophilized
14.4kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Fatty acid binding protein 6, ileal
Assay Standard
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.