missing translation for 'onlineSavingsMsg'
Learn More

F2 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the full length of mouse F2 (NP_034298.1) (Met 1-Gly 618) fused to a C-terminal polyhistidine tag

Brand:  Invitrogen 50344M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15363136

  • £139.50 / 5µg


Estimated Shipment
04-01-2019
Add to basket

Includes: One vial of protein in suspension buffer

Description

Description

Coagulation Factor II (F2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate Boc-VPR-AMC (R&D Systems Cat. No. ES011), the specific activity is >2000pmole/min./μg
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
14061
PAGE
5μg
C-Terminal
Other Proteins|Proteases
RP23-176N9.4, Cf-2, Cf2, FII, thrombin
F2
Proteases, Peptidases & Inhibitors
His Tag
Lyophilized
69.3kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
Coagulation factor II
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.