missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ F11R (JAMA) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 242) of human F11R (NP_058642.1) fused to the fused Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10198H08H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15254895

  • £225.90 / 50µg


Estimated Shipment
09-05-2018
Add to basket

Description

Description

F11 Receptor (F11R), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to support the adhesion of Jurkat human leukemic T cells; when 8 x 104 cells/well are added to F11R coated plates (0.2μg/mL and 100μL/well), approximately 30-40% will adhere specifically after 60 minutes at 37°C
  • Formulation: Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
25kDa
50μg
Viral Host Cell Receptors|Intercellular Adhesion Molecule
JAM, FA11R, KAT, JAM1, JAMA, JCAM, CD321, JAM-1, JAM-A, PAM-1
F11R
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
F11 Receptor
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.