missing translation for 'onlineSavingsMsg'
Learn More

ERBB4 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 651) of human ERBB4 precursor (NP_001036064.1) fused to the His-tagged Fc region of human IgG1 at the C-terminus

Brand:  Invitrogen 10363H03H5

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 13465836

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

V-erb-a erythroblastic leukemia viral oncogene homolog 4 (ERBB4), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human ERBB3-Fc at 1μg/well can bind biotinylated recombinant human NRG1 with a linear range of 0.032-0.16μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
97.5kDa
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
v-erb-a erythroblastic leukemia viral oncogene homolog 4
EGF Receptor (ErbB) Kinase Family
Mammalian
Lyophilized
≥85% by SDS-PAGE
2066
5μg
Truncated/Recombinant/Cell Surface Protein
HER4, mgC138404, p180erbB4, ERBB4
ERBB4
His Tag/IgG1 Fc Tag
Growth Factors & Receptors
Receptor Tyrosine Kinases, Growth Factor Receptors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.