missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ERBB2 Recombinant Mouse Protein (without Catalytic Activity Domain), hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 653) of the mouse ERBB2 (P70424) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50714M02H25

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13453149

  • £138.46 / Pack of 5


Estimated Shipment
16-05-2018
Add to basket

Includes: Five vials of lyophilized protein

Description

Description

V-erb-b2 erythroblastic leukemia viral oncogene homolog 2 (ERBB2), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: 1. Measured by its ability to bind human MUC1 in a functional ELISA; 2. Measured by its ability to bind human CTNNB1 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
96.7kDa
5 x 5μg
Truncated/Recombinant
<1 EU/μg
v-erb-b2 erythroblastic leukemia viral oncogene homolog 2
Assay Standard
Mammalian
IgG1 Fc Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
EGF (Epidermal Growth Factor)|Receptor Tyrosine Kinases
RP23-355L10.8, Erbb-2, HER-2, HER2, Neu, c-erbB2, c-neu, mKIAA3023
ERBB2
Growth Factors & Receptors
EGF Receptor (ErbB) Kinase Family
Videos

For Research Use Only. Not for use in diagnostic procedures.