Learn More
ERBB2 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 652) of human ERBB2 (NP_004439.2) fused to the Fc region of human IgG1 at the C-terminus
Brand: Invitrogen 10004H02H50
Code : NEW
Additional Details : Weight : 0.01000kg
Description
V-erb-b2 erythroblastic leukemia viral oncogene homolog 2 (ERBB2), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.- Activity: Measured by its binding ability in a functional ELISA; immobilized human ERBB2 at 2.5μg/mL can bind Herceptin with a linear range of 3.2-80ng/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
50μg | |
Truncated/Recombinant/Cell Surface Protein | |
ErbB2, NEU, NGL, HER2, TKR1, CD340, HER-2, c-erb B2, HER-2/neu | |
ERBB2 | |
IgG1 Fc Tag | |
Growth Factors & Receptors | |
Receptor Tyrosine Kinases, Growth Factor Receptors |
96.1kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
v-erb-b2 erythroblastic leukemia viral oncogene homolog 2 | |
EGF Receptor (ErbB) Kinase Family | |
Mammalian | |
Lyophilized | |
≥85% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.