Sino Biological™ EPOR Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 250) of human EPOR precursor (NP_000112.1) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10707H08H5

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 13475956

Quantity Price
1 £ 54.75 / 5µg
EU Stock 1
Add to basket


Description and Specification

Specification

Gene Alias EPOR, mgC138358
Structural Form Truncated/Recombinant
Molecular Weight 26.3kDa
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Cytokine Receptors
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Family Hormones & Receptors
Common Name Erythropoietin Receptor
Endotoxin Concentration <1 EU/μg
Gene ID (Entrez) 2057
Protein Tag His Tag
Validated Application Assay Standard
Purity ≥90% by SDS-PAGE
Quantity 5μg
Gene Symbol EPOR
Expression System Mammalian
Format Lyophilized

Erythropoietin receptor (EPOR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CSF2RB-His at 1μg/well can bind biotinylated human EPOR-His with a linear range of 0.16-4.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.