missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EPOR Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met1-Pro 250) of human EPOR (NP_000112.1) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10707H02H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13405956

  • £148.87 / 25µg


Estimated Shipment
09-05-2018
Add to basket

Description

Description

Erythropoietin receptor (EPOR), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized CSF2RB/CD131 at 10μg/mL (100μL/well) can bind biotinylated recombinant human EPOR with a linear range of 0.16-4μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2057
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Cytokine Receptors
EPOR, mgC138358
EPOR
Hormones & Receptors
IgG1 Fc Tag
Lyophilized
51.0kDa
5 x 5μg
Truncated/Recombinant
<1 EU/μg
Erythropoietin Receptor
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.