missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EPOR & CSF2RB (CD131) Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 250) of human EPOR (NP_000112.1) linked by a peptide to the extracellular domain (Trp 17-Trp 443) of human CD131 (NP_000386.1) and fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ CT010H02H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13486426

  • £270.66 / 50µg


Estimated Shipment
01-08-2018
Add to basket

Description

Description

Erythropoietin receptor & colony stimulating factor 2 receptor, beta, low-affinity (EPOR & CSF2RB/ CD131), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2057/1439
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Cytokine Receptors
CD131, EPOR
EPOR CSF2RB
CD Proteins
IgG1 Fc Tag
Lyophilized
125kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Erythropoietin Receptor/Colony stimulating factor 2 receptor, beta, low-affinity
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.