Learn More
EPHA4 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human EPHA4 (NP_031962.2) extracellular domain (Met 1-Thr 547) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus
Brand: Invitrogen 11314H03H5
Additional Details : Weight : 0.10000kg
Description
Eph Receptor A4 (EPHA4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.- Activity: Measured by its binding ability in a functional ELISA; immobilized human EPHA5 at 20μg/mL (100μL/well) can bind human EFNA4-Fc with a linear ranger of 1.28-32ng/mL
- Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Protein Assay Control | |
5μg | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Other Proteins |
For Research Use Only. Not for use in diagnostic procedures.