missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ENO1 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human ENO1 (P06733-1) (Met 1-Lys 434) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 11554H07E250

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218655

  • £454.92 / 250µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

Enolase 1 alpha (ENO1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2023
PAGE
5 x 50μg
Full Length/Recombinant
Enolase 1 Alpha
Assay Standard
E. coli
Frozen
49.3kDa
≥90% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
ENO1L1, MPB1, NNE, PPH
ENO1
Other Proteins
N-His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.