missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ENG Recombinant Human Protein , hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 586) of human ENG (NP_001108225.1) fused to the human IgG1 Fc region at the C-terminus

Brand:  Thermo Scientific™ 10149H02H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15224745

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Endoglin (ENG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind biotinylated human TGFRB1 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
87.5kDa
5 x 5μg
TGFs (Transforming Growth Factors)
RP11-228B15.2, CD105, END, FLJ41744, HHT1, ORW, ORW1
ENG
CD Proteins
IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Endoglin
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.