missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EIF4EBP1 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human 4EBP1 (NP_004086.1) (Ser 2-Ile 118) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10022H07E250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15224525

  • £485.88 / 250µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Eukaryotic Translation Initiation Factor 4E Binding Protein 1 (EIF4EBP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 8.0
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1978
PAGE
5 x 50μg
Full Length/Recombinant
Eukaryotic Translation Initiation Factor 4E Binding Protein 1
Assay Standard
E. coli
Frozen
13.4kDa
≥85% by SDS-PAGE
Store at -5° to -30°C. Store aliquots under sterile conditions at -5° to -30°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
4E-BP1, EIF4EBP1, BP-1, mgC4316, PHAS-I
EIF4EBP1
Other Proteins
N-His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.