missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR Recombinant Mouse Protein (without Catalytic Activity Domain), hIgG1-Fc Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse EGFR (Q01279) (Met 1-Ser 647) fused to the Fc region of human IgG1 at the C-terminus
£57.75 - £250.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molecular Weight (g/mol) | 96kDa |
Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Structural Form | Truncated/Recombinant/Cell Surface Protein |
Endotoxin Concentration | <1 EU/μg |
Includes: Five vials of lyophilized protein
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13473179
|
Invitrogen
51091M02H5 |
5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13493179
|
Invitrogen
51091M02H50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Epidermal Growth Factor Receptor (EGFR), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.- Activity: Measured by its ability to bind human EGF in a functional ELISA
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Epidermal growth factor receptor | |
EGF Receptor (ErbB) Kinase Family | |
Mammalian | |
Lyophilized | |
≥85% by SDS-PAGE |
96kDa | |
Truncated/Recombinant/Cell Surface Protein | |
RP23-295E4.1, 9030024J15Rik, AI552599, Erbb, Errp, Wa5, wa-2, wa2 | |
EGFR | |
IgG1 Fc Tag | |
Growth Factors & Receptors | |
EGF (Epidermal Growth Factor), Receptor Tyrosine Kinases |
For Research Use Only. Not for use in diagnostic procedures.