missing translation for 'onlineSavingsMsg'
Learn More

EGFR Recombinant Mouse Protein (without Catalytic Activity Domain), hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse EGFR (Q01279) (Met 1-Ser 647) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Invitrogen 51091M02H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13463179

  • £887.69 / 250µg


Estimated Shipment
05-03-2019
Add to basket

Includes: Five vials of lyophilized protein

Description

Description

Epidermal Growth Factor Receptor (EGFR), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to bind human EGF in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
96kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Epidermal growth factor receptor
Assay Standard
Mammalian
IgG1 Fc Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
EGF (Epidermal Growth Factor)|Receptor Tyrosine Kinases
RP23-295E4.1, 9030024J15Rik, AI552599, Erbb, Errp, Wa5, wa-2, wa2
EGFR
Growth Factors & Receptors
EGF Receptor (ErbB) Kinase Family
Videos

For Research Use Only. Not for use in diagnostic procedures.