missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EGFR Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 645) of human EGFR (NP_005219) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10001H02H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13412899

  • £55.96 / 5µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Epidermal growth factor receptor (EGFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human EGF at 10μg/mL (100μL/well) can bind human EGFR with a linear range of 0.64-400ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1956
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Receptor Tyrosine Kinases|Growth Factor Receptors
EGF R, ErbB, HER1, mENA, ErbB1, PIG61
EGFR
Growth Factors & Receptors
EGF Receptor (ErbB) Kinase Family
Lyophilized
95kDa
5μg
Truncated/Recombinant
<1 EU/μg
Epidermal growth factor receptor
Assay Standard
Mammalian
IgG1 Fc Tag
Videos