missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EGF Recombinant Mouse Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse EGF (P01132) (Asn 977-Arg 1029) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 50482M01H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13403149

  • £76.26 / 50µg


Estimated Shipment
13-06-2018
Add to basket

Includes: One vial of lyophilized protein

Description

Description

Epidermal growth factor (EGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblasts; the EC50 for this effect is typically 0.2-1ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
34.5kDa
50μg
Full Length/Recombinant
<1 EU/μg
Epidermal growth factor
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
EGF (Epidermal Growth Factor)
AI790464
EGF
Growth Factors & Receptors
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.