Sino Biological™ EFNB2 (EphrinB2) Recombinant Mouse Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 232) of mouse EFNB2 (NP_034241.2) fused to the Fc region of human IgG1 at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 50598M02H5

EFNB2 (EphrinB2) Recombinant Mouse Protein (hIgG1-

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 13436366

Quantity Price
1 £ 54.5 / 5µg
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Specification

Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant/Cell Surface Protein/Ligand
Protein Subtype Ephrins
Gene Alias EFNB2, ELF-2, Epl5, Eplg5, Htk-L, LERK-5, Lerk5, NLERK-1
Gene Symbol EFNB2
Gene ID (Entrez) 13642
Molecular Weight 49kDa
Endotoxin Concentration <1 EU/μg
Validated Application Assay Standard
Protein Family Kinases & Inhibitors
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Common Name Ephrin B2
Protein Tag IgG1 Fc Tag
Expression System Mammalian
Format Lyophilized

Ephrin B2 (EFNB2), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse EPHB4 at 2μg/mL (100μL/well) can bind mouse EFNB2-Fc with a linear range of 0.2-25ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.